Learn More
Biolegend Cyanine3 Donkey anti-rabbit IgG (min. x-reactivity)
Supplier:  Biolegend 406402
Encompass_Preferred
Cyanine3 Donkey anti-rabbit IgG (min. x-reactivity) [Poly4064]; Isotype: Donkey Polyclonal Ig; Reactivity: Rabbit; Apps: ICC; Size: 100 ug
Catalog No. 50-169-142