Learn More
Apexbio Technology LLC Cy3 hydrazide (non-sulfonated) 50mg
SDP
Supplier:  Apexbio Technology LLC A826550
Encompass_Preferred
Cy3 hydrazide (non-sulfonated) is an amine-reactive fluorescent dye with excitation and emission maxima around 550 nm and 570 nm respectively As a hydrazide derivative it specifically interacts with aldehyde or ketone groups introduced onto biomolecules such as oxidized antibodies glycoproteins and oligosaccharides or proteins undergoing carbonylation due to oxidative stress Through covalent linkage formation Cy3 hydrazide fluorescently labels these carbonyl-modified targets facilitating their detection and quantification via fluorescence microscopy electrophoresis or mass spectrometry Its applications include monitoring protein oxidation status glycoconjugates characterization and fluorescent labeling of nucleic acids for biochemical or imaging assays
Catalog No. 50-000-62105