Learn More
Medchemexpress LLC Cy3-N3 | 1621101-43-6 | 93.4% | 712.88 | C34H44N6O7S2 | 10 MG

Supplier: Medchemexpress LLC HYD105110MG
Cy3-N3 is a cyanine-based azide fluorescent dye used to label proteins, peptides, and nucleic acids via click chemistry. The azide functional group enables copper-catalyzed azide-alkyne cycloaddition (CuAAC) and strain-promoted alkyne-azide cycloaddition (SPAAC). Provided as a solid for research use only.
- Azide functional group for CuAAC and SPAAC reactions.
- Suitable for labeling proteins, peptides, and nucleic acids.
- Molecular weight 712.88; formula C34H44N6O7S2.
- Purity approximately 93.4%.
- Supplied as a 10 MG solid; store protected from light.
- Intended for research use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.