Learn More
Medchemexpress LLC Cy3B NHS ester | 228272-52-4 | 97.8% | 10 MG
SDP
Supplier:  Medchemexpress LLC HYD184910MG
Encompass_Preferred
Cy3B NHS ester is a fluorescent labeling reagent that covalently attaches a cyanine-like fluorophore to primary amines via NHS-ester chemistry, enabling bright and photostable labeling of biomolecules for detection and imaging.
  • Reactive N-hydroxysuccinimide (NHS) ester for efficient coupling to primary amines.
  • Bright, photostable cyanine-like fluorophore for fluorescence detection.
  • High purity (97.8%) for reliable labeling performance.
  • Molecular weight 657.73 and CAS number 228272-52-4.
  • Available in multiple package sizes, including 10 mg.
Catalog No. 50-002-97454