Learn More
Medchemexpress LLC Cy5.5 DBCO | 1857352-95-4 | C59H58N4O14S4 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1517205MG
Encompass_Preferred
Cy5.5 DBCO is a click chemistry reagent containing a cycloalkynes group, serving as a linker for the Cyanine5.5 fluorophore. The DBCO group enables copper-free biocompatible click chemistry, characterized by fast reaction kinetics and good stability. It is intended for research use only.
  • Click chemistry reagent.
  • Contains a cycloalkynes group.
  • Functions as a linker for Cyanine5.5 fluorophore.
  • Enables copper-free biocompatible click chemistry.
  • Offers fast reaction kinetics.
  • Provides good stability.
  • Intended for research use only.
Catalog No. 50-003-75008