Learn More
Apexbio Technology LLC Cy5.5 maleimide (non-sulfonated), 5mg. Cas: MFCD: NA. Labeling of cysteine residues, as well as other thiolated molecules
SDP
Supplier:  Apexbio Technology LLC A81405
Encompass_Preferred
Cy5.5 maleimide is a thiol reactive dye which is capable of selective labeling of proteins which contain sulfhydryl groups. Cy5.5 maleimide has near infrared emission that makes it suitable for bioimaging applications. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5055