Learn More
Medchemexpress LLC Cy5.5-PEG-biotin | 10 MG
SDP
Supplier:  Medchemexpress LLC HYD256910MG
Encompass_Preferred
Cy5.5-PEG-Biotin (MW 2000) is a polyethylene glycol derivative conjugated with the Cy5.5 near-infrared fluorophore and a biotin moiety. It is used as a fluorescent labeling and capture reagent for research applications, where biotin enables strong binding to streptavidin/avidin and Cy5.5 provides NIR detection.
  • Cy5.5 fluorophore for near-infrared imaging and detection.
  • PEG spacer (average MW 2000) enhances solubility and reduces steric hindrance.
  • Biotin functional group enables high-affinity capture by streptavidin/avidin.
  • Suitable for labeling proteins, nucleic acids, and surfaces in detection workflows.
  • Supplied as a research reagent, intended for laboratory use.
Catalog No. 50-002-97571