Learn More
Apexbio Technology LLC Cy5 azide (non-sulfonated), 5mg. Cas: MFCD: NA. used for Click chemistry to label alkyne-modified oligonucleotides
SDP
Supplier:  Apexbio Technology LLC A81135
Encompass_Preferred
Cy5 azide is a labeling reagent ready for the use in Click Chemistry reaction which available as 10 mM solution in DMSO. For biomolecule labeling, the labeling reagent has low aqueous solubility, using of organic co-solvent to dissolve this molecular is necessary for efficient reaction. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5011