Learn More
Apexbio Technology LLC Cy5 carboxylic acid, 5mg. Cas: MFCD: NA. Non-reactive fluorophore, for control experiments, and for calibration
SDP
Supplier:  Apexbio Technology LLC A81375
Encompass_Preferred
Sulfo-Cy5 carboxylic acid is a non-activated, sulfonated, hydrophilic and highly water soluble dye. Non-Sulfo-Cy5 carboxylic acid analog is available. For kinds of researches related experiments, using pre-activated sulfo-Cy5 NHS ester is preferred Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5051
 Restricted Item