Learn More
Medchemexpress LLC Cy5-conjugated Goat Anti-Rabbit IgG H&L | 100 UL

Supplier: Medchemexpress LLC HYP81019100UL
Cy5-conjugated Goat Anti-Rabbit IgG H&L is a goat-derived anti-rabbit IgG antibody conjugated with Cy5. It conjugates the light and heavy chains of rabbit IgG antibodies. This product is intended for research use only and not for human use.
- Host: Goat
- Species reactivity: Rabbit
- Cy5 conjugated
- Endogenous sensitivity
- Supplied in 0.01M Phosphate Buffered Saline, pH 7.2, with 1% BSA, 50% glycerol, and 0.02% Sodium Azide
- Stable for 1 year when stored at -20°C
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.