Learn More
Medchemexpress LLC Cy5 maleimide | 1437796-65-0 | 98.0% | 641.24 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY13820050MG
Encompass_Preferred
Cy5 maleimide is a CY dye, a type of Cyanine compound characterized by two nitrogen atoms connected by an odd number of methyl units. These dyes are known for their long wavelength, adjustable absorption and emission, high extinction coefficient, good water solubility, and relatively simple synthesis. It is commonly utilized for the labeling of proteins, antibodies, and small molecular compounds, offering a straightforward mixing reaction for protein antibody labeling.
  • Long wavelength
  • Adjustable absorption and emission
  • High extinction coefficient
  • Good water solubility
  • Relatively simple synthesis
  • Used for labeling proteins
  • Used for labeling antibodies
  • Used for labeling small molecular compounds
Catalog No. 50-003-48175