Learn More
Medchemexpress LLC Cy5 maleimide | 1437796-65-0 | 98.04% | 641.24 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY138200100MG
Encompass_Preferred
Cy5 maleimide is a CY dye characterized by long wavelengths, adjustable absorption and emission, high extinction coefficients, and good water solubility. It is frequently used for labeling proteins, antibodies, and small molecular compounds through a simple mixing reaction.
  • Characterized by long wavelengths
  • Adjustable absorption and emission
  • High extinction coefficients
  • Good water solubility
  • Relatively simple synthesis
  • Used for labeling proteins, antibodies, and small molecular compounds
Catalog No. 50-003-50030