Learn More
Medchemexpress LLC Cy5 maleimide | 1437796-65-0 | 99.4% | C38H45ClN4O3 | 10MG

Supplier: Medchemexpress LLC HY13820010MG
Cy5 maleimide is a thiol-reactive Cyanine5 fluorescent reagent for covalent labeling of cysteine residues in proteins and peptides. It delivers red/NIR fluorescence for imaging and fluorescence-based assays, and is supplied as a brown solid with high purity and recommended cold, light-protected storage.
- High purity (99.43%) ensuring consistent labeling performance.
- Thiol-reactive maleimide group for selective cysteine conjugation.
- Red/NIR excitation and emission (Ex 646 nm, Em 664 nm) for multiplexed assays.
- High solubility in DMSO (≈125 mg/mL) for stock solution preparation.
- Stable when stored sealed at -20°C; in solution keep at -80°C for long-term storage.
- Brown solid appearance for easy handling and verification.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.