Learn More
Medchemexpress LLC Cy5-PEG5-azide bromide | 98.8% | C44H63BrN6O6 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1410591MG
Encompass_Preferred
Cy5-PEG5-azide is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is also a click chemistry reagent containing an Azide group, capable of undergoing copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with Alkyne groups, and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with alkyne groups
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups
Catalog No. 50-003-53135