Learn More
Medchemexpress LLC Cy5-PEG6-NHS ester | 98.5% | C51H71ClN4O11 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1410441MG
Encompass_Preferred
Cy5-PEG6-NHS ester is a PEG-based PROTAC linker designed for research use. It is utilized in the synthesis of PROTACs, which are molecules containing two different ligands connected by a linker. One ligand targets an E3 ubiquitin ligase, and the other targets a specific protein, enabling the selective degradation of target proteins through the intracellular ubiquitin-proteasome system.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Applicable in cancer and cancer targeted therapy research
Catalog No. 50-003-54993