Learn More
Medchemexpress LLC Cy5-UTP (sodium) (10 mM in water) | C45H58N5O22P3S2.xNa | 100 UL
SDP
Supplier:  Medchemexpress LLC HY150145A100UL
Encompass_Preferred
Cy5-UTP sodium (10 mM in water) is a fluorescently labeled ribonucleotide triphosphate that serves as a substrate for terminal deoxynucleotide transferase (TdT) and T7 RNA polymerase. It is utilized to label RNA probes generated in vitro and is applicable in FISH, multi-color fluorescence analysis, and dual-color expression arrays.
  • Fluorescently labeled ribonucleotide triphosphate
  • Substrate for terminal deoxynucleotide transferase (TdT)
  • Substrate for T7 RNA polymerase for in vitro transcription
  • Used to label RNA probes generated in vitro
  • Applicable in FISH and multi-color fluorescence analysis
  • Ideal for dual-color expression arrays
  • Assembled mRNA emits orange fluorescence observable under UV light
Catalog No. 50-003-75003