Learn More
Cayman Chemical Cy5-YntrIethylamIn salt 1mg
SDP
Supplier:  Cayman Chemical 401341 MG
Encompass_Preferred
A clickable fluorescent probe; ex/em = 650/680 nm, respectively; when peptide-conjugated, it has been used as a pH-independent fluorescent probe for the activity of lysosomal cathepsin proteases in S. typhimurium-infected and bystander BMDMs
Catalog No. 50-294-2472