Learn More
Cayman Chemical Cy5trIethylamIn salt 10mg
SDP
Supplier:  Cayman Chemical 3075210 MG
Encompass_Preferred
A cyanine-containing fluorochrome; has commonly been conjugated to secondary antibodies for use in ICC and IHC applications; ex/em = 650/670 nm, respectively
Catalog No. 50-297-8802