Learn More
Selleck Chemical LLC Cy7 E2864-5MG
SDP
Supplier:  Selleck Chemical LLC E28645MG
Encompass_Preferred
Cy7 (Sulfo-Cyanine7) is a fluorescence labelling agent used to label proteins antibodies peptides and oligonucleotides
Catalog No. 50-000-40335