Learn More
Apexbio Technology LLC Cy7.5 carboxylic acid (non-sulfonated), 5mg
SDP
Supplier:  Apexbio Technology LLC A81365
Encompass_Preferred
Cy7.5 carboxylic acid is a free un-activated NIR dye which has low aqueous solubility. As for coupling and labeling reactions, pre-activated Cy7 NHS ester or water-soluble sulfo-Cy7 NHS ester would be considered. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5049