Learn More
Medchemexpress LLC Cy7-conjugated Goat Anti-Mouse IgG H&L | 50 UL
SDP
Supplier:  Medchemexpress LLC HYP8102050UL
Encompass_Preferred
Cy7-conjugated Goat Anti-Mouse IgG H&L is a Cy7-conjugated, goat-derived anti-mouse IgG antibody. It conjugates the light and heavy chains of mouse IgG antibodies and is for research use only.
  • Cy7-conjugated
  • Goat-derived anti-mouse IgG antibody
  • Conjugates light and heavy chains of mouse IgG antibodies
  • For research use only
Catalog No. 50-004-41968