Learn More
Medchemexpress LLC HY-14264 5mg Medchemexpress, Cyamemazine CAS:3546-03-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY142645MG
Encompass_Preferred
Medchemexpress, HY-14264 5mg Cyamemazine CAS:3546-03-0 Cyamemazine is a neuroleptic agent that contains the phenothiazine chromophore. Cyamemazine is often used as an anxiolytic. Cyamemazine is a potent 5-HT3 (Ki of 12 nM), 5-HT2A (Ki = 1.5 nM) and 5-HT2C (Ki of 75 nM) receptors antagonist with antipsychotic activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9895