Learn More
Enzo Life Sciences Cyanidin chloride (10mg). CAS: 528-58-5
SDP
Supplier:  Enzo Life Sciences ALX385003M010
Encompass_Preferred
Antioxidant flavonoid. Nitric oxide (NO) scavenger. Alternative name: 3,3',4',5,7-Pentahydroxy-flavylium chloride. Purity: ≥97% (HPLC). Appearance: Red to black solid. Solubility: Soluble in methanol or 0.1% hydrochloric acid. Long Term Storage: -20°C.
Catalog No. 50-200-9035