Learn More
STA PHARMACEUTICAL US LLC Cyanine 3 amidite | 1 g | InChIKey GTLRVZNKJSCYOV-UHFFFAOYSA-M | MW 1045.100231998
Supplier: STA PHARMACEUTICAL US LLC STAAM10517MM301GM
Cyanine 3 amidite is a Amidite reagent (Subcategory: Dye Amidite) sold by WuXi TIDES. Offered in 1 g. Store at -20 °C. Ships on dry ice. SDS available for reference.
Specifications
- CAS: No data
- MDL: No data
- InChIKey: GTLRVZNKJSCYOV-UHFFFAOYSA-M
- Molecular Weight: 1045.100231998
- Molecular Formula: C58H70IN4O4P
- Purity: ≥85%
- Container Type: 30 mL Glass (28-400)
- Pack Size: 1 g
- Net Weight: 1 g
- Gross Weight: 50.1 g
- Commodity Code: 29339980
- Country Of Origin: China
- IUPAC: 2-((E)-3-((Z)-1-(3-(((2-cyanoethoxy)(diisopropylamino)phosphaneyl)oxy)propyl)-3,3-dimethylindolin-2-ylidene)prop-1-en-1-yl)-1-(3-((4-methoxyphenyl)diphenylmethoxy)propyl)-3,3-dimethyl-3H-indol-1-ium iodide
- SMILES: CC(N(P(OCCCN1/C(C(C)(C2=C1C=CC=C2)C)=C\C=C\C3=[N+](C4=C(C3(C)C)C=CC=C4)CCCOC(C5=CC=CC=C5)(C6=CC=C(C=C6)OC)C7=CC=CC=C7)OCCC#N)C(C)C)C.[I-]
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.