Learn More
Enzo Life Sciences Cyanine 5 UTP enhanced (250 nmol)
SDP
Supplier:  Enzo Life Sciences ENZ42506
Encompass_Preferred
Cyanine 5-UTP (Enhanced) can replace UTP as a substrate for T7 RNA polymerase in labeling systems that generate labeled probes through in vitro transcription. Probes generated by these methods are suitable for multicolor fluorescence analysis, specifically dual-color expression arrays in conjunction with cyanine 3-UTP. Alternative Name: Cyanine 5-uridine-5'-triphosphate (enhanced). Formulation: Liquid. Solution in water. Purity: ≥93% (HPLC). Appearance: Blue liquid. Use/Stability: Stable for at least one year after receipt when stored as recommended. Shipping: Shipped on Dry Ice. Long Term Storage: -20°C Excitation maximum: 650 nm Emission maximum: 665 nm. Handling: Protect from light. Avoid freeze/thaw cycles.
Catalog No. 50-103-1809