Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cyanine3 azide chloride | 1167421-28-4 | 98.9% | 575.19 | 1 MG
SDP

Supplier:  Medchemexpress LLC HYD13271MG

Encompass_Preferred

Cyanine3 azide chloride is an analog of Cy3 azide and functions as a potent green fluorescent dye. It is utilized in click chemistry for coupling with alkyne-labeled proteins. As a click chemistry reagent, it contains an azide group, enabling it to participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules. It can also undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.

  • Potent green fluorescent dye.
  • Uses click chemistry for coupling with alkyne-labeled proteins.
  • Detectable by fluorometers, imagers, and microscopes.
  • Contains an azide group for copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions.
  • Can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions.
  • Excitation (Ex) at 555 nm and Emission (Em) at 569 nm.

Catalog No. 50-004-32013


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.