Learn More
Medchemexpress LLC Cyanine3 azide chloride | 1167421-28-4 | MFCD00006236 | 99.4% | 575.19 g/mol | C33H43ClN6O | 10 MG
SDP
Supplier:  Medchemexpress LLC HYD132710MG
Encompass_Preferred
Cyanine3 azide chloride is a Cy3-range fluorescent azide reagent designed for covalent labeling of biomolecules via copper-catalyzed azide-alkyne click chemistry. Supplied as a solid (C33H43ClN6O, MW 575.19 g/mol) with high purity, it produces bright fluorescence suitable for detection and imaging applications.
  • Provides bright fluorescence with excitation at 555 nm and emission at 569 nm.
  • Compatible with copper-catalyzed azide-alkyne cycloaddition (CuAAC) for efficient biomolecule labeling.
  • Supplied as a solid with high purity (99.4%) for reliable experimental results.
  • Available in small research pack sizes, including 10 mg, for convenient use.
  • Suitable for fluorescence detection and imaging of labeled proteins, nucleic acids, and other biomolecules.
Catalog No. 50-002-97445