Learn More
Medchemexpress LLC Cyanine5.5 alkyne chloride | 1628790-37-3 | 99.0% | 656.30 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYD13255MG
Encompass_Preferred
Cyanine5.5 alkyne chloride is an alkynyl-modified near-infrared (NIR) fluorescent dye (Ex=678 nm, Em=694 nm). It can react with azides for click chemistry labeling.
  • Near-infrared (NIR) fluorescent dye
  • Suitable for click chemistry labeling
Catalog No. 50-004-32012