Learn More
Medchemexpress LLC Cyanine5 azide chloride | 1267539-32-1 | 99.6% | 601.22 g·mol⁻¹ | C35H45ClN6O | 10 MG
SDP
Supplier:  Medchemexpress LLC HYD132010MG
Encompass_Preferred
Cyanine5 azide chloride is a near-infrared fluorescent labeling reagent containing an azide functional group for click chemistry. It is used to conjugate to alkyne-modified biomolecules for fluorescence imaging and probe synthesis.
  • Azide functional group enables copper-catalyzed and strain-promoted click reactions.
  • Near-infrared excitation ≈646-647 nm and emission ≈662-668 nm for low background imaging.
  • High purity (≈99.6%) suitable for labeling applications.
  • Soluble in DMSO at 100 mg/mL for convenient stock solutions.
  • Available in small laboratory pack sizes for research use.
Catalog No. 50-002-97081