Learn More
Medchemexpress LLC α-cyano-4-hydroxycinnamic acid | 28166-41-8 | 99.9% | 189.17 | C10H7NO3 | 250 MG
SDP
Supplier:  Medchemexpress LLC HY107641250MG
Encompass_Preferred
α-Cyano-4-hydroxycinnamic acid is a cinnamic acid derivative used in research as an inhibitor of monocarboxylate transporters and as a matrix for MALDI-TOF mass spectrometry peptide ionization. It also inhibits mitochondrial pyruvate transport (Ki = 6.3 μM) and is supplied as a high-purity reagent with defined storage recommendations for powder and solutions.
  • Potent inhibitor of monocarboxylate transporters and mitochondrial pyruvate transport (Ki = 6.3 μM).
  • Commonly used as a MALDI-TOF matrix to facilitate peptide ionization.
  • High purity suitable for analytical and biological research (reported purity ~99.9%).
  • Available as solids and as prepared solutions for experimental convenience.
  • Stable when stored under recommended conditions for powder and solvent forms.
Catalog No. 50-002-40038