Learn More
Medchemexpress LLC β-cyano-L-Alanine | 6232-19-5 | 114.10 | 50 MG
SDP
Supplier:  Medchemexpress LLC 50MGHY125773
Encompass_Preferred
β-cyano-L-Alanine is a nitrile commonly found in higher plants, enzymatically produced by cyanoalanine synthase from cyanide and cysteine. It has been observed to abolish the protective effect of ethanol on cerebral ischemia/reperfusion (I/R) injury.
  • Can be used in research related to neurological diseases
  • Functions as an endogenous metabolite
  • Soluble in water and PBS for various experimental setups
  • Can be stored long-term at -20°C (under nitrogen) or -80°C in solvent
Catalog No. 50-003-66130
 Restricted Item