Learn More
Medchemexpress LLC Cyanocobalamin (Vitamin B12 Standard) | 68-19-9 | 99.95% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB0315R100MG
Encompass_Preferred
Vitamin B12 (Standard) is the analytical standard of Vitamin B12. This product is intended for research and analytical applications. Vitamin B12 plays a key role in the normal functioning of the brain and nervous system, and for the formation of blood.
  • Analytical standard for research and analytical applications
  • Supports normal functioning of the brain and nervous system
  • Essential for the formation of blood
  • Suitable as a reference standard in HPLC, GC, and MS experiments
Catalog No. 50-004-06690