Learn More
Cayman Chemical cyanoLAlanine Inhibitors
SDP
Supplier:  Cayman Chemical 1001094750 MG
Encompass_Preferred
A reversible inhibitor of the H2S synthesizing enzyme cystathionine-γ-lyase; blocks H2S synthesis in rat liver preparations with an IC50 value of 6.5 µM
Catalog No. 50-288-9230