Learn More
Medchemexpress LLC Cyanox CY 1790 500g | 40601-76-1 | 699.90 g·mol⁻¹ | C42H57N3O6 | 500 G
SDP
Supplier:  Medchemexpress LLC HY116772500G
Encompass_Preferred
Cyanox 1790 is a powdered hindered phenolic antioxidant (CAS 40601-76-1) used to provide long-term thermal and processing stabilization for polyolefins, polyamides, styrenics, and other polymers in extrusion and molding applications.
  • Provides long-term thermal and processing stabilization for polymers.
  • Acts as an effective free-radical scavenger to protect against degradation.
  • Has low volatility and low color contribution for color-sensitive formulations.
  • Resists gas-fade and discoloration during extrusion and heat exposure.
  • Typical effective loading is about 0.05-0.08% by weight in polymer formulations.
Catalog No. 50-003-08026