Learn More
Medchemexpress LLC Cyanox CY 1790 50g | 40601-76-1 | 50 G
SDP
Supplier:  Medchemexpress LLC HY11677250G
Encompass_Preferred
Cyanox CY 1790 is a sterically hindered phenolic antioxidant used to protect polyethylene and other polymers from thermal and oxidative degradation. It exhibits resistance to chlorine dioxide and is supplied as a solid suitable for research and formulation work.
  • Sterically hindered phenolic antioxidant for polymer stabilization.
  • Resistant to chlorine dioxide-induced degradation.
  • Solid form; soluble in DMSO (≥ 100 mg/mL) for formulation or analysis.
  • Molecular weight approximately 699.92 g·mol-1; formula C42H57N3O6.
  • Intended for research and anti-degradation studies of polyethylene.
Catalog No. 50-002-27187