Learn More
Medchemexpress LLC Cyantraniliprole-d3 (HGW-86-d3) | 1392493-34-3 | 99.6% | 476.73 g/mol | C19H11D3BrClN6O2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY12779S5MG
Encompass_Preferred
Cyantraniliprole-d3 is the deuterium-labeled analog of the insecticide cyantraniliprole, provided as an isotopically labeled reference standard for analytical and research applications. It is intended for research use only and is supplied as a solid (white to off-white).
  • Deuterium-labeled analog for use as an internal standard in LC-MS/MS quantitation
  • High purity suitable for analytical workflows
  • Available in small, pre-weighed quantities for method development
  • Solid, white to off-white appearance for easy visual inspection
  • Useful for pesticide residue analysis and method validation
Catalog No. 50-002-64604