Learn More
ANALYTIK JENA US LLC Analytik Jena CyBio RoboTipTray 384, PCR-Certified, Pre-Sterilized, 60 µL
Supplier:  ANALYTIK JENA US LLC OL381025244
Encompass
The Analytik Jena CyBio RoboTipTray 384/60 µL is a premium consumable for automated liquid handling, featuring 384 pipette tips per tray to maximize throughput and efficiency. Designed with PCR-certified, pre-sterilized tips from high-grade virgin polypropylene, it ensures precise, low-retention transfers, making it ideal for high-throughput applications. Its compatibility with CyBio FeliX liquid handlers sets it apart, promoting accuracy in pipetting across various plate formats, enhancing laboratory productivity.
Catalog No. NC1376150