Learn More
Medchemexpress LLC Cyclanoline chloride | 17472-50-3 | MFCD00221739 | 96.3% | 377.86 | C20H24ClNO4 | 5 MG

Supplier: Medchemexpress LLC HY1206925MG
Cyclanoline chloride is the chloride salt of cyclanoline, a small-molecule cholinesterase inhibitor supplied for research use as a solid reagent for in vitro biochemical and cell-based studies.
- Cholinesterase inhibitory activity demonstrated in biochemical assays.
- Molecular formula C20H24ClNO4 and molecular weight 377.86.
- Solid, off-white to light brown appearance.
- Soluble in DMSO at 100 mg/mL; use newly opened DMSO and ultrasonic agitation if needed.
- Research-grade purity reported at approximately 96.3%.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.