Learn More
Medchemexpress LLC Cyclen | 294-90-6 | 172.28 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW007656100G
Encompass_Preferred
Cyclen is a macrocyclic tetraamine chelating agent and the aza analogue of crown ether. It is utilized as a precursor for MRI contrast agents and as an intermediate for preparing effective macrocyclic chelates. It functions as a structural regulator in the interfacial polymerization of polyethyleneimine (PEI) and trimesoyl chloride (TMC) to develop polyamide NF membranes for efficient Li+/Mg2+ separation. It possesses a specific cavity structure and exhibits selective coordination properties for Li+ ions.
  • Macrocyclic tetraamine chelating agent
  • Aza analogue of crown ether
  • Utilized as a precursor for MRI contrast agents
  • Intermediate for preparing effective macrocyclic chelates
  • Functions as a structural regulator in interfacial polymerization
  • Possesses a specific cavity structure
  • Exhibits selective coordination properties for Li+ ions
Catalog No. 50-004-46551