Learn More
Medchemexpress LLC Cyclic adenosine monophosphate-13C5 | 2170095-23-3 | 97.45% | 500 UG
SDP
Supplier:  Medchemexpress LLC HYB1511S500UG
Encompass_Preferred
Cyclic AMP-13C5 is a deuterated cyclic AMP. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Deuterated cyclic AMP.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotope.
  • Potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-11959