Learn More
Medchemexpress LLC HY-B1511 500mg Medchemexpress, Cyclic AMP CAS:60-92-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB1511500MG
Encompass_Preferred
Medchemexpress, HY-B1511 500mg Cyclic AMP CAS:60-92-4 Cyclic AMP (cAMP) is a mitogenic messenger and promotes the G 1 to S phase transition in the cell cycle. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2959