Learn More
Medchemexpress LLC Cyclic nona-L-arginine (hydrochloride) | >98.4% | 1442.13 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYP3193A1MG
Encompass_Preferred
Cyclic nona-L-arginine hydrochloride is a nonaarginine peptide primarily utilized as a laboratory chemical for drug delivery studies. It demonstrates faster translocation compared to its linear counterparts and is intended for research use only.
  • Nonaarginine peptide
  • Utilized for drug delivery
  • Demonstrates faster translocation
  • Solid, white to off-white appearance
  • Soluble in water (100 mg/mL with sonication)
  • Long-term stability when stored as powder at -80°C or -20°C
  • Stable in solvent for shorter periods when stored at -80°C or -20°C
Catalog No. 50-003-97218