Learn More
Abcam Cyclic pifithrin-alpha-p-nitro, transcriptional p53 inhibitor, 5MG
Supplier:  Abcam AB1460305MG
Encompass_Preferred
MW 299.3 Da, Purity >99%. Cell-permeable transcriptional p53 inhibitor. Shows potent neuroprotective effects (ED₅₀ = 30 nM, etoposide-induced cortical neuron death). Shows a 50% longer half-life than Pifithrin-α (ab120478).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4957