Learn More
Selleck Chemical LLC Cyclizine hydrochloride E4905-100MG
SDP
Supplier:  Selleck Chemical LLC E4905100MG
Encompass_Preferred
Cyclizine hydrochloride is an antagonist of the H1 histamine receptor used for the treatment and prevention of vertigo and motion sickness It also exhibits antihistaminic properties
Catalog No. 50-000-41870