Learn More
CPC Scientific Cyclo(-Gly-Arg-Gly-Asp-Ser-Pro) (trifluoroacetate salt) 10MG
SDP
Supplier:  CPC Scientific RGDP001C
Encompass
This cyclic peptide is a potent vasodilator. It is more powerful than the linear GRGDSP in changing the vascular tone of arterioles isolated from rat cremaster muscleSEQUENCE: Cyclo(-Gly-Arg-Gly-Asp-Ser-Pro) (trifluoroacetate salt)ONE-LETTER SEQUENCE: GRGDSP, N to C cyclizedMOLECULAR FORMULA: C22H35N9O9MOLECULAR WEIGHT: 569.57STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [135432-37-0]SYNONYMS: c(GRGDSP)RESEARCH AREA: CancerREFERENCES:Mogford, JE. et al. Circulation Res. 79, 821 (1996)SKU(s): RGDP-001A, RGDP-001B, RGDP-001C, RGDP-001D
Catalog No. 50-211-0371