Learn More
CPC Scientific Cyclo(-Arg-Gly-Asp-D-Phe-Lys) (trifluoroacetate salt) 25MG
SDP
Supplier:  CPC Scientific RGDP004C
Encompass
In one study where this peptide was labeled with 125I, it was found to bind specifically and with high affinity to alphav3 receptors on neovascular blood vessel sections of different major human cancers. The integrin alpha(IIb)beta(3)-specific cyclic hexapeptide contains an Arg-Gly-Asp (RGD) sequence.SEQUENCE: Cyclo(-Arg-Gly-Asp-D-Phe-Lys) (trifluoroacetate salt)ONE-LETTER SEQUENCE: Cyclo(-RGDfK)MOLECULAR FORMULA: C27H41N9O7MOLECULAR WEIGHT: 603.67STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [161552-03-0]SYNONYMS: c(RGDfK)RESEARCH AREA: CancerREFERENCES:Van Hagen, PM. et al. Int. J. Cancer 90, 186 (2000)Hu, B. et al. Biochem. 39, 2284 (2000).M. Kantlehner, et al., Chembiochem., 1, 107 (2000).R.J. Kok, et al., Bioconjug. Chem., 13, 128 (2002).SKU(s): RGDP-004A, RGDP-004B, RGDP-004C
Catalog No. 50-211-0380