Learn More
Carbosynth LLC. Cyclo(Arg-Ala-Asp-D-Phe-Lys), 5mg, MW: (M.W. 617.71), Molecular formula: C28H43N9O7, CAS# [756500-23-9]
SDP
Supplier:  Carbosynth LLC. PCI3883PI5MG
Encompass_Preferred
Synonyms: cyclo (Arg-Ala-Asp-D-Phe-Lys), c(RADfK), Application: Negative Control RGD Peptide for PCI-3661-PI, Storage Temperature: -20°C, References: P.K. Dubey, V. Mishra, S. Jain, S. Mahor, and S.P. Vyas, J. Drug Targeting, 12, 257 (2004).
Catalog No. 50-168-6309