Learn More
Medchemexpress LLC Cyclo(Arg-Gly-Asp-D-Phe-Cys) (Cyclo(RGDfC)) | 862772-11-0 | 98.1% | 578.64 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP23005MG
Encompass_Preferred
Cyclo(Arg-Gly-Asp-D-Phe-Cys) (Cyclo RGDfC) is a cyclic RGD peptide with high affinity to αvβ3, capable of disrupting cell integrin interactions. It inhibits pluripotent marker expression in embryonic stem cells (ESCs) and the tumorigenic potential of mESCs in vivo.
  • High affinity to αvβ3
  • Disrupts cell integrin interactions
  • Inhibits pluripotent marker expression in embryonic stem cells
  • Inhibits tumorigenic potential of mESCs in vivo
  • Down-regulates transcription factors Oct 4, Sox 2, and Nanog in mESCs
  • Inhibits integrin gene expression in mESC-col I constructs
  • Promotes aggregate formation and detachment in mESCs
  • Applicable in tumor research
Catalog No. 50-004-23918