Learn More
Carbosynth LLC. Cyclo (Arg-Gly-Asp-D-Phe-Cys), 1mg, MW: (M.W. 578.65), Molecular formula: C24H34N8O7S, CAS# [862772-11-0]
SDP
Supplier:  Carbosynth LLC. PCI3686PI1MG
Encompass_Preferred
This item has a minimum order quantity of 3 per supplier requirements
Synonyms: cyclo (Arg-Gly-Asp-D-Phe-Cys), c(RGDfC), Application: RGD Tumor Targeting Peptide (linker additions via Cys) (May require further derivatization before use), Salt Form:, Acetate Salt, Storage Temperature: -20C, References: C. Pattillo, F. Sari-Sarraf, R. Nallamothu, B. Moore, G. Wood, and M. Kiani, Experim. Clin.Therapeutics, Radiation Research Society Meeting (2005)
Catalog No. 50-168-6279