Learn More
Medchemexpress LLC Cyclo(Arg-Gly-Asp-(D-Phe)-Glu) | 756500-22-8 | 99.94% | 604.61 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP50215MG
Encompass_Preferred
Cyclo(Arg-Gly-Asp-(D-Phe)-Glu) c(RGDfE) is a cyclic RGD peptide targeting integrin αvβ3. It is commonly used for modifying drug-loaded nanoparticles and is often utilized in cancer research, such as pancreatic cancer.
  • Targets integrin αvβ3.
  • Used for modifying drug-loaded nanoparticles.
  • Utilized in cancer research.
  • Enhances targeting ability of nanoparticles towards BxPC3 cells.
  • Enhances targeting of nanoparticles towards integrin αvβ3 expressing cells.
Catalog No. 50-003-98425